Assay Details
Gene Symbol: PLPP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023638254.1 | XP_023494022.1 | 182 | 74 | ||
XM_023638263.1 | XP_023494031.1 | 190 | 74 | |||
XM_023638264.1 | XP_023494032.1 | 189 | 74 | |||
XM_023638272.1 | XP_023494040.1 | 199 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
