Assay Details
Gene Symbol: PDZD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023638712.1 | XP_023494480.1 | 1319 | 92 | ||
XM_023638713.1 | XP_023494481.1 | 1193 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() |
