Assay Details
Gene Symbol: STN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005602258.3 | XP_005602315.1 | 915 | 98 | ||
XM_005602259.3 | XP_005602316.1 | 709 | 98 | |||
XM_014733314.2 | XP_014588800.1 | 988 | 98 | |||
XM_023639977.1 | XP_023495745.1 | 915 | 98 | |||
XM_023639983.1 | XP_023495751.1 | 1836 | 98 | |||
XM_023639987.1 | XP_023495755.1 | 916 | 98 | |||
XM_023639997.1 | XP_023495765.1 | 1772 | 98 | |||
XM_023640004.1 | XP_023495772.1 | 916 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
