Assay Details
Gene Symbol: NEURL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023640053.1 | XP_023495821.1 | 1746 | 49 | ||
XM_023640062.1 | XP_023495830.1 | 1426 | 49 |

More Information
Set Membership: |
![]() ![]() ![]() |
