Assay Details
Gene Symbol: CNNM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023640147.1 | XP_023495915.1 | 2406 | 65 | ||
XM_023640152.1 | XP_023495920.1 | 2486 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
