Assay Details
Gene Symbol: BORCS7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001499572.6 | XP_001499622.2 | 751 | 92 | ||
XM_014733297.2 | XP_014588783.1 | 787 | 92 | |||
GenBank mRNA | JL615992.1 | - | 1 - 2 | 188 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
