Assay Details
Gene Symbol: WBP1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001498544.5 | XP_001498594.1 | 177 | 95 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: WBP1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001498544.5 | XP_001498594.1 | 177 | 95 |