Assay Details
Gene Symbol: WBP1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001498544.5 | XP_001498594.1 | 434 | 91 | ||
XM_005602268.3 | XP_005602325.1 | 450 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() |
