Assay Details
Gene Symbol: MFSD13A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001498826.5 | XP_001498876.1 | 989 | 63 | ||
XM_014733346.2 | XP_014588832.1 | 817 | 63 | |||
XM_023640281.1 | XP_023496049.1 | 989 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
