Assay Details
Gene Symbol: ELOVL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001499143.5 | XP_001499193.3 | 1350 | 72 | ||
XM_005602276.3 | XP_005602333.1 | 184 | 72 | |||
XM_023640430.1 | XP_023496198.1 | 650 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
