Assay Details
Gene Symbol: KCNIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001499376.6 | XP_001499426.2 | 863 | 59 | ||
XM_001499403.4 | XP_001499453.1 | 1065 | 59 | |||
XM_001499409.4 | XP_001499459.2 | 4178 | 59 | |||
XM_001499421.4 | XP_001499471.1 | 703 | 59 | |||
XM_014733041.2 | XP_014588527.1 | 865 | 59 | |||
XM_023640574.1 | XP_023496342.1 | 1021 | 59 | |||
XM_023640575.1 | XP_023496343.1 | 1018 | 59 | |||
XM_023640580.1 | XP_023496348.1 | 965 | 59 | |||
XM_023640586.1 | XP_023496354.1 | 962 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
