Assay Details
Gene Symbol: OGA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001499535.5 | XP_001499585.1 | 1631 | 84 | ||
XM_023640514.1 | XP_023496282.1 | 1280 | 84 | |||
XM_023640519.1 | XP_023496287.1 | 2900 | 84 |
