Assay Details
Gene Symbol: DPCD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001499801.5 | XP_001499851.1 | 594 | 59 | ||
XM_014733061.2 | XP_014588547.1 | 476 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
