Assay Details
Gene Symbol: SLF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023640832.1 | XP_023496600.1 | 3766 | 108 | ||
XM_023640834.1 | XP_023496602.1 | 3762 | 108 | |||
XM_023640837.1 | XP_023496605.1 | 3680 | 108 | |||
XM_023640839.1 | XP_023496607.1 | 4046 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
