Assay Details
Gene Symbol: HPS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023641202.1 | XP_023496970.1 | 1769 | 58 | ||
XM_023641203.1 | XP_023496971.1 | 2438 | 58 | |||
XM_023641213.1 | XP_023496981.1 | 1656 | 58 | |||
XM_023641219.1 | XP_023496987.1 | 2442 | 58 | |||
XM_023641224.1 | XP_023496992.1 | 1617 | 58 | |||
XM_023641231.1 | XP_023496999.1 | 2433 | 58 | |||
XM_023641238.1 | XP_023497006.1 | 1373 | 58 | |||
XM_023641240.1 | XP_023497008.1 | 1246 | 58 | |||
XM_023641245.1 | XP_023497013.1 | 1206 | 58 | |||
XM_023641253.1 | XP_023497021.1 | 2699 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() |
