Assay Details
Gene Symbol: OPALIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005602350.3 | XP_005602407.1 | 644 | 80 | ||
XM_023641753.1 | XP_023497521.1 | 607 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
