Assay Details
Gene Symbol: LGI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001502400.5 | XP_001502450.3 | 823 | 82 | ||
XM_023642325.1 | XP_023498093.1 | 752 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
