Assay Details
Gene Symbol: CPEB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014733057.2 | XP_014588543.2 | 3038 | 69 | ||
XM_023642591.1 | XP_023498359.1 | 1720 | 69 | |||
XM_023642610.1 | XP_023498378.1 | 1654 | 69 | |||
XM_023642617.1 | XP_023498385.1 | 1696 | 69 | |||
XM_023642619.1 | XP_023498387.1 | 1651 | 69 | |||
XM_023642624.1 | XP_023498392.1 | 1627 | 69 | |||
XM_023642627.1 | XP_023498395.1 | 1722 | 69 | |||
XM_023642638.1 | XP_023498406.1 | 1722 | 69 | |||
XM_023642644.1 | XP_023498412.1 | 1721 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
