Assay Details
Gene Symbol: ACTA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001503035.6 | XP_001503085.1 | 150 | 86 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001503035.6 transcripts.
Set Membership: |
![]() ![]() |
