Assay Details
Gene Symbol: LIPJ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001501539.4 | XP_001501589.3 | 1639 | 132 | ||
XM_023643145.1 | XP_023498913.1 | 1765 | 132 | |||
XM_023643148.1 | XP_023498916.1 | 1201 | 132 | |||
XM_023643149.1 | XP_023498917.1 | 1864 | 132 | |||
XM_023643155.1 | XP_023498923.1 | 1766 | 132 |

More Information
Set Membership: |
![]() ![]() ![]() |
