Assay Details
Gene Symbol: LIPJ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001501539.4 | XP_001501589.3 | 1779 | 89 | ||
XM_023643145.1 | XP_023498913.1 | 1905 | 89 | |||
XM_023643148.1 | XP_023498916.1 | 1341 | 89 | |||
XM_023643149.1 | XP_023498917.1 | 2004 | 89 | |||
XM_023643160.1 | XP_023498928.1 | 1759 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
