Assay Details
Gene Symbol: IPMK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023643438.1 | XP_023499206.1 | 890 | 100 | ||
XM_023643441.1 | XP_023499209.1 | 880 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() |
