Assay Details
Gene Symbol: TFAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001503382.4 | XP_001503432.1 | 713 | 79 | ||
XM_023643450.1 | XP_023499218.1 | 602 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() |
