Assay Details
Gene Symbol: PHYHIPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001502093.5 | XP_001502143.2 | 1436 | 68 | ||
XM_014733317.2 | XP_014588803.1 | 1870 | 68 | |||
XM_023643534.1 | XP_023499302.1 | 842 | 68 | |||
XM_023643549.1 | XP_023499317.1 | 1973 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
