Assay Details
Gene Symbol: PCBD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014733392.2 | XP_014588878.1 | 628 | 63 | ||
XM_023644471.1 | XP_023500239.1 | 305 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
