Assay Details
Gene Symbol: PLA2G12B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001503808.6 | XP_001503858.3 | 389 | 96 | ||
XM_005602461.3 | XP_005602518.1 | 389 | 96 | |||
XM_014733394.2 | XP_014588880.1 | 389 | 96 | |||
XM_023644891.1 | XP_023500659.1 | 389 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
