Assay Details
Gene Symbol: PLA2G12B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001503808.6 | XP_001503858.3 | 475 | 61 | ||
XM_014733394.2 | XP_014588880.1 | 475 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
