Assay Details
Gene Symbol: NUDT13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001503824.4 | XP_001503874.1 | 1068 | 65 | ||
XM_005602462.3 | XP_005602519.1 | 1181 | 65 | |||
XM_014733037.2 | XP_014588523.1 | 1079 | 65 | |||
XM_014733038.2 | XP_014588524.1 | 522 | 65 | |||
XM_023644987.1 | XP_023500755.1 | 1221 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
