Assay Details
Gene Symbol: MSS51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014733357.2 | XP_014588843.1 | 2837 | 74 | ||
XM_014733358.2 | XP_014588844.1 | 505 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
