Assay Details
Gene Symbol: PGBD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023646259.1 | XP_023502027.1 | 1295 | 62 | ||
XM_023646260.1 | XP_023502028.1 | 1295 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
