Assay Details
Gene Symbol: RAB4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001497955.6 | XP_001498005.2 | 565 | 92 | ||
XM_014733855.2 | XP_014589341.1 | 272 | 92 | |||
XM_023646351.1 | XP_023502119.1 | 565 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
