Assay Details
Gene Symbol: CXCL12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001489644.5 | XP_001489694.1 | 274 | 83 | ||
XM_005602634.3 | XP_005602691.1 | 274 | 83 | |||
XM_005602636.3 | XP_005602693.1 | 274 | 83 | |||
GenBank mRNA | KF612401.1 | - | 2 - 3 | 178 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
