Assay Details
Gene Symbol: ERO1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001492252.6 | XP_001492302.2 | 880 | 73 | ||
XM_023647026.1 | XP_023502794.1 | 880 | 73 | |||
XM_023647037.1 | XP_023502805.1 | 880 | 73 | |||
XM_023647040.1 | XP_023502808.1 | 880 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
