Assay Details
Gene Symbol: GPR137B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014733713.2 | XP_014589199.2 | 1158 | 58 | ||
XM_023647052.1 | XP_023502820.1 | 1154 | 58 | |||
XM_023647062.1 | XP_023502830.1 | 1042 | 58 | |||
XM_023647070.1 | XP_023502838.1 | 1039 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
