Assay Details
Gene Symbol: MAP3K21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023647429.1 | XP_023503197.1 | 1845 | 79 | ||
XM_023647436.1 | XP_023503204.1 | 2024 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() |
