Assay Details
Gene Symbol: ARV1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023647787.1 | XP_023503555.1 | 749 | 90 | ||
XM_023647795.1 | XP_023503563.1 | 749 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
