Assay Details
Gene Symbol: PTPN20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005602729.3 | XP_005602786.1 | 1466 | 93 | ||
XM_005602732.3 | XP_005602789.1 | 1336 | 93 | |||
XM_014734101.2 | XP_014589587.1 | 1319 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() |
