Assay Details
Gene Symbol: PTPN20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005602729.3 | XP_005602786.1 | 410 | 56 | ||
XM_005602732.3 | XP_005602789.1 | 410 | 56 | |||
XM_014734101.2 | XP_014589587.1 | 410 | 56 | |||
XM_023648426.1 | XP_023504194.1 | 411 | 56 | |||
XM_023648429.1 | XP_023504197.1 | 414 | 56 | |||
XM_023648433.1 | XP_023504201.1 | 419 | 56 | |||
XM_023648436.1 | XP_023504204.1 | 394 | 56 | |||
XM_023648440.1 | XP_023504208.1 | 401 | 56 | |||
XM_023648447.1 | XP_023504215.1 | 320 | 56 | |||
XM_023648449.1 | XP_023504217.1 | 331 | 56 | |||
XM_023648455.1 | XP_023504223.1 | 325 | 56 | |||
XM_023648464.1 | XP_023504232.1 | 411 | 56 | |||
XM_023648478.1 | XP_023504246.1 | 410 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() |
