Assay Details
Gene Symbol: PRXL2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005602747.3 | XP_005602804.1 | 411 | 101 | ||
XM_005602748.3 | XP_005602805.1 | 318 | 101 | |||
XM_014733888.2 | XP_014589374.1 | 429 | 101 | |||
XM_023648961.1 | XP_023504729.1 | 401 | 101 | |||
XM_023648969.1 | XP_023504737.1 | 405 | 101 |

More Information
Set Membership: |
![]() ![]() |
