Assay Details
Gene Symbol: DYDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001501456.4 | XP_001501506.1 | 574 | 67 | ||
XM_023648982.1 | XP_023504750.1 | 571 | 67 | |||
XM_023648987.1 | XP_023504755.1 | 456 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
