Assay Details
Gene Symbol: BLM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005602774.3 | XP_005602831.1 | 245 | 79 | ||
XM_023649579.1 | XP_023505347.1 | 1343 | 79 | |||
XM_023649584.1 | XP_023505352.1 | 1334 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() |
