Assay Details
Gene Symbol: POLR3GL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023641307.1 | XP_023497075.1 | 664 | 75 | ||
XM_023641308.1 | XP_023497076.1 | 517 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |
