Assay Details
Gene Symbol: CHRNA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023652702.1 | XP_023508470.1 | 624 | 65 | ||
XM_023652709.1 | XP_023508477.1 | 505 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
