Assay Details
Gene Symbol: CHRNB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014734089.2 | XP_014589575.2 | 1592 | 67 | ||
XM_014734090.2 | XP_014589576.2 | 1490 | 67 | |||
XM_023652754.1 | XP_023508522.1 | 2955 | 67 | |||
XM_023652769.1 | XP_023508537.1 | 1388 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
