Assay Details
Gene Symbol: COX5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023653440.1 | XP_023509208.1 | 469 | 92 | ||
GenBank mRNA | JL632690.1 | - | 2 - 3 | 519 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() |
