Assay Details
Gene Symbol: CYP1A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001493886.5 | XP_001493936.3 | 1239 | 67 | ||
XM_014734103.2 | XP_014589589.2 | 1190 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
