Assay Details
Gene Symbol: STRA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023653756.1 | XP_023509524.1 | 1925 | 64 | ||
XM_023653757.1 | XP_023509525.1 | 1843 | 64 | |||
XM_023653759.1 | XP_023509527.1 | 2092 | 64 | |||
XM_023653760.1 | XP_023509528.1 | 2062 | 64 | |||
XM_023653761.1 | XP_023509529.1 | 2031 | 64 | |||
XM_023653762.1 | XP_023509530.1 | 2250 | 64 | |||
XM_023653763.1 | XP_023509531.1 | 2029 | 64 | |||
XM_023653764.1 | XP_023509532.1 | 1998 | 64 | |||
XM_023653767.1 | XP_023509535.1 | 2148 | 64 | |||
XM_023653769.1 | XP_023509537.1 | 2109 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
