Assay Details
Gene Symbol: PML | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001493244.4 | XP_001493294.1 | 1520 | 75 | ||
XM_005602932.3 | XP_005602989.2 | 1520 | 75 | |||
XM_005602934.3 | XP_005602991.2 | 1520 | 75 | |||
XM_005602935.3 | XP_005602992.1 | 1520 | 75 | |||
XM_014733729.2 | XP_014589215.1 | 1519 | 75 | |||
XM_014733730.2 | XP_014589216.1 | 1519 | 75 | |||
XM_014733731.2 | XP_014589217.1 | 1519 | 75 | |||
XM_023653830.1 | XP_023509598.1 | 1521 | 75 | |||
XM_023653834.1 | XP_023509602.1 | 1520 | 75 | |||
XM_023653841.1 | XP_023509609.1 | 1520 | 75 | |||
XM_023653849.1 | XP_023509617.1 | 1520 | 75 | |||
XM_023653859.1 | XP_023509627.1 | 1520 | 75 | |||
XM_023653863.1 | XP_023509631.1 | 1520 | 75 | |||
XM_023653867.1 | XP_023509635.1 | 1520 | 75 | |||
XM_023653875.1 | XP_023509643.1 | 1520 | 75 | |||
XM_023653885.1 | XP_023509653.1 | 1520 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
