Assay Details
Gene Symbol: ADPGK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001493993.5 | XP_001494043.3 | 534 | 64 | ||
XM_005602948.3 | XP_005603005.2 | 534 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
