Assay Details
Gene Symbol: GLCE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023654752.1 | XP_023510520.1 | 876 | 66 | ||
XM_023654759.1 | XP_023510527.1 | 788 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
